Learn More
ATAGO™ Digital Hand-Held Pocket Honey Refractometer, PAL-22S
Measures the % water content of honey easily and displays readings digitally. The Atago Pal-22S Digital Hand-Held Pocket Honey Refractometer is comfortable, easy to use, lightweight and compact.
Brand: ATAGO™ 4422
Code : A0
Additional Details : Weight : 0.10000kg
Description
- Measurements can be taken anywhere
Order Info
Carrying case (PAL-CASE) is not included
Specifications
Refractometer | |
PAL-22S | |
0.10% | |
10°C to 40°C |
12.0 to 30.0% | |
±0.2 % | |
Honey Moisture |