Learn More
Attune™ NxT Emission Filter, 620/15BP
Attune NxT Emission Filter, 620/15BP
Brand: Invitrogen 100022762
Additional Details : Weight : 0.10000kg
Description
This is a replacement 620/15 bandpass filter used in the optics path of the Attune™ NxT Acoustic Focusing Cytometer.Specifications
Attune Acoustic Focusing Cytometer | |
Room temperature | |
Attune™ NxT Emission Filter | |
1 Each |
For Research Use Only. Not for use in diagnostic procedures.