missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Attune™ NxT Emission Filter, 710/50BP
This is a replacement 710/50 bandpass filter used in the optics path of the Attune™ NxT Acoustic Focusing Cytometer.
Brand: Invitrogen 100022765
Additional Details : Weight : 0.10000kg
Description
- Replacement bandpass filter
- Used in the optics path of the Attune™ NxT Acoustic Focusing Cytometer
Specifications
Attune Acoustic Focusing Cytometer | |
Store at room temperature. | |
Emission Filter | |
Attune™ |
For Research Use Only. Not for use in diagnostic procedures.