Learn More
Attune™ Wash Solution
Brand:  Invitrogen A24974
42.86 EUR valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Additional Details : Weight : 0.30000kg
 View more versions of this product
Product Code. 15360445
  • 45.12 € / 250mL
In Stock
to see stock.
Description
Description
Attune™ Wash Solution is an optimized cleaning buffer that is used in the Attune™ Acoustic Focusing Cytometer to maintain cleanliness of the fluidics lines, minimizing contamination or carryover.
Maintains System Cleanliness
Attune™ Wash Solution has been formulated to remove cellular debris, prevent protein building up, and minimize dyes sticking to the fluidics lines of the system. It is recommended for use during the routine maintenance and automated shutdown procedures of the Attune™ Acoustic Focusing Cytometer.
Convenient
The ready-to-use 1X wash solution was designed to eliminate the need for dilution prior to use, as well as access to a purified water system. The wash solution is stable for 30 days after the bottle is opened.
High Quality
The Attune™ Wash Solution is prepared, filtered, and packaged in a sterile environment to ensure the highest quality. Each batch is traceable by its lot number designation and has been validated for use on the Attune™ Acoustic Focusing Cytometer.
Specifications
Specifications
Store at room temperature.
250 mL
Attune™
Wash Solution
Attune™ Acoustic Focusing Cytometer