Learn More
Labcaire™ AU-1222-ETHE ETHE Fume Cabinet Filter
Brand:  Labcaire™ AU-1222
Code : 04
Additional Details : Weight : 12.00000kg
Product Code. 12881770
  • 1075.90 € / Each
Estimated Shipment: 15-07-2024
to see stock.
Specifications
ETHE
12 Kg
ETHE
Main For Diethyl/Methyl Ether
Aura Model 250/550
Fume Cabinet Filter
AU-1222-ETHE