Learn More
Carl Zeiss™ AxioVision Rel. 4.8.2 Software
Control high-sensitivity microscopes, and capture, process, annotate, and catalog images with this software download. Carl Zeiss™ AxioVision Rel. 4.8.2 Software helps you perform complex and minute tasks simply and intuitively.
Brand: Carl Zeiss™ 410130-0600-000
Additional Details : Weight : 0.80000kg
Description
- Images can be acquired using any common camera with TWAIN driver
- Open and compare up to 21 different image file formats including multidimensional ZVI-images
- Adjust display curve, brightness, contrast and color balance
- Acquire length, contour and angle of microscopical structures. Export the results to other programs
- Apply comments and scale bars to your image
- Print directly from AxioVision LE or export your image to various other file formats
Specifications
AxioVision Rel. 4.8.2 Software | |
Light microscopes, PC | |
1 Ea. |