missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ 0.5 mL Self Standing Screw Cap Tubes
Features O-ring manufactured for resistant to many commonly-used laboratory chemicals
158.00 € - 1895.80 €
Specifications
Max. RCF | 20,000 x g |
---|---|
Certifications/Compliance | RNase, DNase, pyrogen-free |
Closure Material | Polypropylene |
Capacity (Metric) | 0.5 mL |
Material | Polypropylene |
Product Code | Brand | Closure Color | Sterility | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Color | Sterility | Price | Quantity & Availability | ||||
11340355
|
Axygen™
SCT-050-SS-A-S |
Assorted | Sterile |
269.00 €
Pack of 500 |
Estimated Shipment: 09-10-2024
Log in to see stock. |
||||
11370355
|
Axygen™
SCT-050-SS-C |
Clear | Non-sterile |
158.00 €
Pack of 500 |
Estimated Shipment: 27-09-2024
Log in to see stock. |
||||
12709508
|
Axygen™
SCT-050-SS-BR-S |
Brown | Sterile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12799498
|
Axygen™
SCT-050-SS-BR |
Brown | Non-sterile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
13194624
|
Axygen™
SCT-050-SS-A |
Assorted | Non-sterile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12739508
|
Axygen™
SCT-050-SS-C-S |
Clear | Sterile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
13161784
|
Axygen™
SCT-050-SS-G |
Green | Non-sterile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12779498
|
Axygen™
SCT-050-SS-A-S |
Assorted | Sterile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12759508
|
Axygen™
SCT-050-SS-GR |
Gray | Non-sterile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10567270
|
Axygen™
SCT-050-SS-GR-S |
Gray | Sterile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10337839
|
Axygen™
SCT-050-SS-G-S |
Green | Sterile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
13167394
|
Axygen™
SCT-050-SS-B |
Blue | Non-sterile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12719518
|
Axygen™
SCT-050-SS-R |
Red | Non-sterile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10312250
|
Axygen™
SCT-050-SS-R-S |
Red | Sterile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
13151784
|
Axygen™
SCT-050-SS-C |
Clear | Non-sterile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12749518
|
Axygen™
SCT-050-SS-V-S |
Violet | Sterile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12739518
|
Axygen™
SCT-050-SS-V |
Violet | Non-sterile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12759518
|
Axygen™
SCT-050-SS-W |
White | Non-sterile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10713392
|
Axygen™
SCT-050-SS-Y-S |
Yellow | Sterile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12789518
|
Axygen™
SCT-050-SS-Y |
Yellow | Non-sterile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
For a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping.
- Available as presterilized and nonsterile tubes
- Nonsterile cap tubes have unattached, non-looped caps with O-rings
- Presterilized cap tubes have non-looped caps attached with O-rings
- Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
- Maxymum Recovery™ screw cap tubes available as special order items for assorted tubes
- Wide range of colored tubes for easy sample identification
Specifications
20,000 x g | |
Polypropylene | |
Polypropylene | |
O-ring | |
Conical |
RNase, DNase, pyrogen-free | |
0.5 mL | |
Non-looped | |
Screw Cap |