Learn More
Axygen™ 1.5 mL Conical Screw Cap Tubes
With chemical-resistant O-ring
Brand:  Axygen™ SCT-150-W
Code : 66
Additional Details : Weight : 7.30000kg
 View more versions of this product
Product Code. 12769558
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping.
  • Available as presterilized and nonsterile
  • Nonsterile cap tubes have unattached, non-looped caps with O-rings
  • Presterilized cap tubes have non-looped caps attached with O-rings
  • Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
  • Maxymum Recovery™ screw cap tubes available as special order
  • Caps available in range of colors for easy sample identification
Specifications
Specifications
20,000 x g
White
1.5 mL
Non-looped
Non-sterile
Screw Cap
RNase, DNase, pyrogen-free
Polypropylene
Polypropylene
O-ring
No
Conical