missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Black Hanging Format Automation Pipet Tips for Hamilton Microlab™ Star Line and Nimbus™, 5 Pack
Compatible with Hamilton Microlab™ Star Line and Nimbus™ automated liquid handling workstations
441.00 € - 920.00 €
Specifications
Color | Black |
---|---|
Autoclavable | Not Autoclavable |
Clean Claims | DNase, RNase and Pyrogen free |
For Use With (Application) | Available in four sizes and specially designed for use with Hamilton™ liquid handling platforms, they enable precision pipetting across a wide range of applications |
Format | Hanging Tip Rack |
Product Code | Brand | Volume (Metric) | Sterility | Filtered | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Sterility | Filtered | Price | Quantity & Availability | |||
15551393
|
Axygen™
HT-10-CBK-HTR-S-5 |
10 μL | Sterile | Non-Filtered |
785.00 €
Pack of 5760 |
Estimated Shipment: 12-07-2024
Log in to see stock. |
|||
15541393
|
Axygen™
HT-10-CBK-HTR-5 |
10 μL | Non-sterile | Non-Filtered |
720.00 €
Pack of 5760 |
Estimated Shipment: 12-07-2024
Log in to see stock. |
|||
15501413
|
Axygen™
HTF-10-CBK-HTR-S-5 |
10 μL | Sterile | Filtered |
920.00 €
Pack of 5760 |
Estimated Shipment: 05-07-2024
Log in to see stock. |
|||
15571403
|
Axygen™
HT-50-CBK-HTR-5 |
50 μL | Non-sterile | Non-Filtered |
665.00 €
Pack of 5760 |
Estimated Shipment: 12-07-2024
Log in to see stock. |
|||
15581403
|
Axygen™
HT-50-CBK-HTR-S-5 |
50 μL | Sterile | Non-Filtered |
725.00 €
Pack of 5760 |
Estimated Shipment: 12-07-2024
Log in to see stock. |
|||
15561413
|
Axygen™
HTF-50-CBK-HTR-S-5 |
50 μL | Sterile | Filtered |
855.00 €
Pack of 5760 |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
15531403
|
Axygen™
HT-300-CBK-HTR-S-5 |
300 μL | Sterile | Non-Filtered |
725.00 €
Pack of 5760 |
Estimated Shipment: 12-07-2024
Log in to see stock. |
|||
15581393
|
Axygen™
HT-1000-CBK-HTR-5 |
1000 μL | Non-sterile | Non-Filtered |
441.00 €
Pack of 3840 |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
15521413
|
Axygen™
HTF-1000-CBKHTRS5 |
1000 μL | Sterile | Filtered |
570.00 €
Pack of 3840 |
Estimated Shipment: 05-07-2024
Log in to see stock. |
|||
15541413
|
Axygen™
HTF-300-CBK-HTRS5 |
300 μL | Sterile | Filtered |
855.00 €
Pack of 5760 |
Estimated Shipment: 05-07-2024
Log in to see stock. |
|||
Description
Axygen offers an alternative choice for your Hamilton™ STARline system.
These tips are:
- Manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality
- Available in non-sterile, sterile, nonfiltered, and filtered
- Available as Clear or Conductive black polypropylene tips
- Available in 96 well hanging tip rack, nested tip rack, and individual deck-seating rack format
- Designed for Hamilton™ MICROLAB™ STAR, STARlet, STARplus and Nimbus™ workstations
Specifications
Black | |
DNase, RNase and Pyrogen free | |
Hanging Tip Rack |
Not Autoclavable | |
Available in four sizes and specially designed for use with Hamilton™ liquid handling platforms, they enable precision pipetting across a wide range of applications | |
Polypropylene |