Learn More
Axygen™ PlateMax™ Semi-Automated Plate Sealer Accessory, Heat Sealing Films
Designed for applications ranging from PCR and Real-Time PCR to ELISA and cell culture
Brand:  Axygen™ HS-100-QPCR
Code : 66
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 12769618
  • 585.00 € / Pack of 500
In Stock
to see stock.
Description
Description
Axygen PlateMax microplate heat sealing films are designed for applications ranging from PCR and Real-time PCR to storage. They can be used in a wide range of temperatures making them suitable for nearly any application.
Specifications
Specifications
Heat Sealing Film for qPCR
PlateMax Plate Sealer
500/Cs.
Plate Sealing
Polyester
100/Pk.