missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Presterilized Conical Centrifuge Tubes

Graduated tubes with conical bottom

Brand:  Axygen™ SCT-50ML-25-S

159.30 EUR valid until 2024-12-31
Use promo code "23272" to get your promotional price.



Code : 66

Additional Details : Weight : 11.34000kg

 View more versions of this product

Product Code. 12519107

  • 174.00 € / Pack of 500
In Stock
to see stock.
Add to basket

Description

Description

  • Rated to 12,000 RCF
  • Printed graduations and writing area for sample identification
  • Screw caps can be closed with one hand.
  • Sterile options are also available.
  • RNase-/DNase-free
  • Nonpyrogenic
Specifications

Specifications

12,000 x g
Not Autoclavable
Clear
For sample identification
25 Tubes and caps/Unit; 20 Units/Cs.
Conical
Screw Cap
50 mL
Polypropylene
Screw cap
Sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers