missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azadibenzocyclooctyne-PEG4-NHS ester, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Description
Azadibenzocyclooctyne-PEG4-NHS ester is applied for protein-peptide conjugation, peptide-small molecule conjugation, 18F radiolabeling, protein-oligonucleotide conjugation and surface modification.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Azadibenzocyclooctyne-PEG4-NHS ester is applied for protein-peptide conjugation, peptide-small molecule conjugation, 18F radiolabeling, protein-oligonucleotide conjugation and surface modification.
Solubility
Soluble in DCM, DMF, DMSO and THF.
Notes
Moisture sensitive. Incompatible with oxidizing agents and heat.
Azadibenzocyclooctyne-PEG4-NHS ester is applied for protein-peptide conjugation, peptide-small molecule conjugation, 18F radiolabeling, protein-oligonucleotide conjugation and surface modification.
Solubility
Soluble in DCM, DMF, DMSO and THF.
Notes
Moisture sensitive. Incompatible with oxidizing agents and heat.
Specifications
Color | Orange to Brown |
Quantity | 10 mg |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in DCM,DMF,DMSO and THF. |
Formula Weight | 649.69 |
Physical Form | Oil |
Chemical Name or Material | Azadibenzocyclooctyne-PEG4-NHS ester |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'