missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AZD 8055

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
AZD 8055 is a highly potent, selective and ATP-competitive mTOR inhibitor (IC50 = 0.8 nM). Inhibits growth and induces autophagy in a range of cancer cell lines and xenografts, including NSCLC and glioblastoma (IC50 values are 20, 50 and 53 nM in H838, A549 and U87MG cell lines, respectively). Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 5-[2,4-Bis[(3S)-3-methyl-4-morpholinyl]pyrido[2,3-d]pyrimidin-7-yl]-2-methoxybenzenemethanol |
| CAS | 1009298-09-2 |
| Quantity | 50 mg |
| Target | mTOR Inhibitors |
| Molecular Formula | C25H31N5O4 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction