missing translation for 'onlineSavingsMsg'
Learn More

Azidotrimethylsilane, 94%, Thermo Scientific Chemicals

Catalog Number 10429270
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
4648-54-8
C3H9N3Si
115.21
MFCD00001986
SEDZOYHHAIAQIW-UHFFFAOYSA-N
trimethylsilyl azide, silane, azidotrimethyl, trimethylsilylazide, azido trimethyl silane, ccris 8048, tms azide, tms-azide, trimethyl silylazide, trimethylsilyl-azide, azido trimethylsilane
78378
azido(trimethyl)silane
C[Si](C)(C)N=[N+]=[N-]
4648-54-8
C3H9N3Si
115.21
MFCD00001986
SEDZOYHHAIAQIW-UHFFFAOYSA-N
trimethylsilyl azide, silane, azidotrimethyl, trimethylsilylazide, azido trimethyl silane, ccris 8048, tms azide, tms-azide, trimethyl silylazide, trimethylsilyl-azide, azido trimethylsilane
78378
azido(trimethyl)silane
C[Si](C)(C)N=[N+]=[N-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, For Azidation/Diazo transfer
TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 0.8760g/mL
Boiling Point 95.0°C to 96.0°C
Flash Point 6°C
Infrared Spectrum Authentic
Assay Percent Range 92% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3SiN3
Refractive Index 1.4142 to 1.4162
Quantity 10 mL
Fieser 01,1236; 03,316; 05,719; 06,632; 07,394; 09,21; 10,14; 11,32; 12,37; 13,24; 14,25; 15,16; 16,17; 17,378
Specific Gravity 0.876
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in ether, dichloromethane and benzene
Formula Weight 115.21
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material Azidotrimethylsilane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Azidotrimethylsilane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Flammable liquid Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H225-Highly flammable liquid and vapour.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'