missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ Lid for Square Shaped HDPE Bucket

For use with square shaped HDPE bucket

13.75 €

Specifications

Product Type Lid
Material HDPE
Autoclavable No
For Use With (Equipment) Square Bucket
Height (Metric) 45 mm
View More Specs

Products 2
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
12085562
View Documents Promotion Details
Azlon
BWX118
White This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12045562
View Documents Promotion Details
Azlon
BWX110
Black This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

  • Lids with moulded inset handle for square shaped buckets
Specifications

Specifications

Lid
No
45 mm
1 Ea.
HDPE
Square Bucket
280 mm
280 mm
Videos
SDS
Documents

Documents

Product Certifications