Learn More
Azlon™ Lightweight Polypropylene Funnels
Lightweight polypropylene made from food grade plastic
7.90 € - 56.80 €
Product Code | Brand | Diameter (Metric) Stem | Height (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Stem | Height (Metric) | Price | Quantity & Availability | ||||
12024608
|
Azlon
FWC060 |
10mm | 107 mm |
26.70 €
Pack of 10 |
Estimated Shipment: 22-04-2024
Log in to see stock. |
||||
12501456
|
Azlon
FWC062 |
13mm | 149 mm |
56.80 €
Pack of 10 |
Estimated Shipment: 22-04-2024
Log in to see stock. |
||||
12551506
|
Azlon
FWC064 |
14mm | 170 mm |
30.00 €
Pack of 5 |
Estimated Shipment: 24-04-2024
Log in to see stock. |
||||
12044598
|
Azlon
FWC066 |
17mm | 210 mm |
42.50 €
Pack of 5 |
Estimated Shipment: 22-04-2024
Log in to see stock. |
||||
12591446
|
Azlon
FWC058 |
9mm | 78 mm |
17.05 €
Pack of 10 |
Estimated Shipment: 22-04-2024
Log in to see stock. |
||||
12581446
|
Azlon
FWC056 |
5mm | 62 mm |
20.70 €
Pack of 10 |
Estimated Shipment: 29-04-2024
Log in to see stock. |
||||
10708205
|
Azlon
FWC054 |
5mm | 47 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Manufactured from lightweight polypropylene. Azlon™ Lightweight Polypropylene Funnels include ribs that prevent airlocks and allow rapid filtration. Autoclavable to 121 °C.
- Lightweight polypropylene funnels with ribs that prevent air locks, allowing rapid filtration
- Autoclavable to 121°C
- Made from food grade plastic
Specifications
Polypropylene | |
Yes |