missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azlon™ Single Piece Safety Shield
For protection in potentially hazardous situations
Brand: Azlon SGT102
Code : NEW
Additional Details : Weight : 3.90000kg
Description
- Incredibly strong and lightweight
- Manufactured from single piece of 4mm thick sheet of transparent polycarbonate
- Supplied with two pairs of plastic coated metal feet
- Feet may be clamped or bolted onto bench to give stability
Specifications
Two pairs of plastic coated metal feet | |
178 mm | |
Polycarbonate | |
230 mm (Center Panel), 178 mm (Outside Panel) |
Shield | |
1 | |
608 mm |