Learn More
Azlon™ Polypropylene Round Scoops
Sturdy scoops in rigid white polypropylene.
Brand: Azlon SWL202
Additional Details : Weight : 0.09450kg
Description
- Suitable for food contact applications in accordance with U.S. FDA regulations
- Round design facilitates use when withdrawing substances from bags and sacks
Specifications
Round Scoop | |
100 mL | |
130 mm |
Polypropylene | |
White |