Learn More
Azlon™ PTFE Stirrer Bars
Ideal for use with large vessels.
46.05 € - 113.00 €
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
15778087
|
Azlon
SWN677 |
75 mm | 13 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15788087
|
Azlon
SWN680 |
108 mm | 27 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15798087
|
Azlon
SWN682 |
159 mm | 27 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Similar to cylindrical stirrer bars but give greater turbulence at low speed
Specifications
Magnet | |
2 |