Learn More
Gibco™ Bacto™ Malt Extract
The water-soluble portion of malted barley.
89.75 €
Description
- Bacto Malt Extract is the water-soluble portion of malted barley
- Due to the extraction process, this animal origin-free supplement provides a very high carbohydrate content to any medium
- It has been used successfully in microbial fermentation applications
- Store in cool, dry conditions in well-sealed containers at 2°C to 25°C
Specifications
Powder | |
Animal origin-free, malted barley |