missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Bacto™ TC Lactalbumin Hydrolysate
An animal origin peptone from the enzymatically hydrolyzed protein portion of milk whey.
POA
Specifications
Content And Storage | Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers. |
---|---|
Form | Powder |
Product Type | Dehydrated Culture Media |
For Use With (Application) | Mammalian, bacterial, insect cell types, recombinant proteins and animal health vaccines |
Product Line | Bacto™ |
Description
This supplement is a mixture of peptides, amino acids, and carbohydrates, both simple and complex. Customers have successfully used Gibco™ Bacto™ TC Lactalbumin Hydrolysate to support the growth of many mammalian, bacterial, and insect cell types, and it has been used to produce both recombinant proteins and animal health vaccines. Customers have also used this supplement as an amino acid supplement for mammalian cell culture media.Specifications
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers. | |
Dehydrated Culture Media | |
Bacto™ |
Powder | |
Mammalian, bacterial, insect cell types, recombinant proteins and animal health vaccines | |
Animal Origin |