missing translation for 'onlineSavingsMsg'
Learn More

Barium Carbonate, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog Number p-5390699
Click to view available options
Quantity:
500 g
Packaging:
Wide Mouth Amber Glass Bottle
This item is not returnable. View return policy
513-77-9
CBaO3
197.34
MFCD00003448
AYJRCSIUFZENHW-UHFFFAOYSA-L
barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p
10563
[Ba++].[O-]C([O-])=O
This item is not returnable. View return policy
513-77-9
CBaO3
197.34
MFCD00003448
AYJRCSIUFZENHW-UHFFFAOYSA-L
barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p
10563
[Ba++].[O-]C([O-])=O

High quality chemicals for laboratory and specialized industrial use

Physical Form Powder
Quantity 500 g
IUPAC Name barium(2+);carbonate
Formula Weight 197.34
Grade BAKER ANALYZED™ ACS Reagent
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 4.30kg
Chemical Name or Material Barium Carbonate
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P330-Rinse mouth.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'