missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium carbonate, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
500 g
Description
Barium carbonate is frequently employed in ceramic industry as a component of glazes. It is added to clay to precipitate some soluble salts while manufacturing bricks, tiles and potteries. Barium carbonate is mainly used as a raw material in the electrical engineering industries for glass production, and as a raw material in the magnet manufacturing industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | (decomposition) |
Melting Point | 811°C |
Physical Form | Lumps, Flakes and Granular Powder |
Quantity | 25 g |
Merck Index | 14,969 |
Solubility Information | Insoluble in water. Soluble in most of the acids,with the exception of sulfuric acid. |
IUPAC Name | barium(2+) carbonate |
Formula Weight | 197.35 |
Percent Purity | 99.997% |
Grade | Puratronic™ |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'