missing translation for 'onlineSavingsMsg'
Learn More

Barium carbonate, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog Number p-45777
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
513-77-9
CBaO3
197.34
MFCD00003448
AYJRCSIUFZENHW-UHFFFAOYSA-L
barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p
10563
[Ba++].[O-]C([O-])=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
513-77-9
CBaO3
197.34
MFCD00003448
AYJRCSIUFZENHW-UHFFFAOYSA-L
barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p
10563
[Ba++].[O-]C([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 99+%
Linear Formula BaCO3
Merck Index 15, 965
IUPAC Name barium(2+);carbonate
Formula Weight 197.35
Percent Purity 99.0 to 101.0%
Grade ACS Reagent
Packaging Plastic bottle
Density 4.4300g/mL
Chemical Name or Material Barium carbonate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'