missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium Hydroxide Octahydrate, Certified AR for Analysis, Fisher Chemical™
Click to view available options
Quantity:
500 g
1 kg
Description
H2BaO2.8H2O, CAS Number-12230-71-6, 500g, White, 315.48g/mol, 78 deg.C, 149152, Plastic powder jar, 12.5, Solid
Specifications
Melting Point | 78°C |
Color | White |
pH | 12.5 |
Physical Form | Solid |
Quantity | 500 g |
Formula Weight | 315.48g/mol |
Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Barium hydroxide octahydrate
- Danger
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 A
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H314-Causes severe skin burns and eye damage.
- EUH071-Corrosive to the respiratory tract.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'