missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Barnstead™ Easypure™ Cartridges and Filters

Product Code. 11426348 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
EasyPure II RF
Easypure
Easypure II
Easypure II UV, Easypure II UV/UF
Easypure II, Easypure UF
Easypure UF, Easypure II UV/UF
Type:
Cartridge Pack, Deionized Feed
Cartridge Pack, RO or Distilled Feed
Cartridge and Filter
Mixed Bed COOL Cartridge
Sanitation Cartridge
Ultrafilter
Resin Type:
2/3 Macroreticular Resin
Mixed Bed
Unit Size:
Each
Filter Size:
6 in.
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Type Resin Type unitSize Filter Size
11426348 Easypure II Cartridge and Filter 2/3 Macroreticular Resin Each -
10422962 Easypure II UV, Easypure II UV/UF Cartridge Pack, Deionized Feed - Each -
10895960 Easypure Mixed Bed COOL Cartridge Mixed Bed Each 6 in.
10728154 Easypure II UV, Easypure II UV/UF Cartridge Pack, RO or Distilled Feed - Each -
10041672 Easypure II, Easypure UF Cartridge Pack, Deionized Feed - Each -
10133502 Easypure II, Easypure UF Cartridge Pack, RO or Distilled Feed - Each -
10498453 Easypure UF, Easypure II UV/UF Ultrafilter - Each -
10502074 EasyPure II RF Sanitation Cartridge - Each -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 11426348 Lieferant Thermo Scientific™ Lieferanten-Nr. D50230

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Extend the life of your Thermo Scientific™ Barnstead™ Easypure™ II or Easypure™ RF water system with replacement cartridges and filters.

Extend the life of your Thermo Scientific™ Barnstead™ Easypure™ II or Easypure™ RF water system with Barnstead™ Easypure™ Cartridges and Filters. Choose the cartridge pack according to your feed water and system.

  • Easypure™ II Systems require pretreated feedwater
  • From the chart, select appropriate cartridges based on application and feedwater type

Bestellinformation

Cartridges and filters must be ordered separately

TRUSTED_SUSTAINABILITY

Spezifikation

Feedwater Source Deionized
Purification Method Activated Carbon
Resin Type 2/3 Macroreticular Resin
For Use With (Application) Organic/colloidal removal
For Use With (Equipment) Easypure II
Type Cartridge and Filter
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.