Learn More
Thermo Scientific™ Barnstead™ Point of Use Water Purification Systems Ultrafilters
Lengthen the life of your water purification system with these replacement ultrafilters, which are automatically flushed.
Brand:  Thermo Scientific™ 50133981
Code : 51
Additional Details : Weight : 0.01800kg
 View more versions of this product
Product Code. 11317171
  • 446.00 € / Each
Estimated Shipment: 28-06-2024
to see stock.
Add to basket
Description
Description
Lengthen the life of your water purification system with replacement Thermo Scientific™ Barnstead™ Point-of-Use Water Purification Systems Ultrafilters. For use with Barnstead point-of-use water purification systems, these internal ultrafilters are flushed automatically to allow for a long two-year lifetime.
Use with the following Barnstead point-of-use water purification systems:
Part 50133980:
  • GenPure™†
  • GenPure™ Pro†
  • GenPure™ xCAD†
†UF, UF/UV and UF/UV-TOC units only
Part 50133981:
  • MicroPure™‡
  • Smart2 Pure™‡
‡UF and UF/UV units only
Specifications
Specifications
Ultrafilter; For use with MicroPure, Smart2 Pure (UF and UF/UV units only)
MicroPure, Smart2Pure (UF and UF/UV units only)
Ultrafilter