Learn More
Thermo Scientific™ Barnstead™ TII Type 2 Water System
This item is no longer available and has been replaced by the next generation of Barnstead Water Purification Systems.
5805.62 €
Specifications
Bacterial retention | 99% |
---|---|
Certifications/Compliance | CE, CSA |
Conductivity | 0.067 to 0.1 µS/cm |
Bacterial Content | <1 CFU/mL |
Feed Water Monitoring | No |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10464913
|
Thermo Scientific™
D14031 |
Discontinued; replacement product is 50132131 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
This item is no longer available and has been replaced by the next generation of Barnstead Water Purification Systems. The recommended replacement for this product is the Thermo Scientific™ Barnstead™ Pacific™ TII water purification system.
Features:
- Green/yellow/red indicator lights simplify operation with detailed status on alert information.
- System is available in 12L and 24L/hr. production rates with 30L or 60L storage tanks for a variety of application needs.
- System monitors water at all points including feedwater, RO water, final product water.
- System measures resistivity and temperature.
- Removable display can be mounted up to 10′ (3m) from unit for added flexibility.
- Space-saving design: unit can be wall-mounted or placed on benchtop UV lamp status can be monitored on display to ensure bacterial-free, low-organic water.
- Cartridge disconnects easily for simple, fast replacement.
- Fluorinated PE reservoir eliminates leaching of organic materials and protects against contamination.
- Direct dispensing offers options of volumetric (0.25 to 60L) or timed dispense.
- Multi-step purification combines pretreatment, reverse osmosis, deionization, UV oxidation and filtration in one.
- System easily dispenses from unit or reservoir.
- Exclusive flow pattern and unique water storage and recirculation system continuously maintain stored water.
- Smart controls monitor critical system data and automate operation, ensuring maximum water quality.
- Sophisticated multifunctional display provides real-time information on water quality, system performance and diagnostics.
- Intuitive multifunction main menu has push-button controls to monitor critical system data.
- System allows easy access to alarm information, setup data and real-time operational readings such as water purity, percent RO rejection, cartridge status and reservoir levels.
Ordering Information:
Optional accessories include remote dispensers, printer, cutoff float, interlock cable, storage reservoirs, bottles and replacement consumables. These products are available separately. For details, contact your sales representative.
Includes:
- Wall bracket
- RO membrane(s)
- UV lamp
Requires:
- Start-Up Kit (D502138)
- Storage reservoir
Recommended for:
- Media prep
- Buffer preps
- Glassware washing
- Equipment feed
- Type 1 Water system feed
Specifications
99% | |
0.067 to 0.1 µS/cm | |
No | |
Type 2 | |
No | |
Water System | |
4°C to 40°C | |
1 L/min. | |
13.5 in. | |
19.5 in. | |
18.5 in. | |
100/240 VAC 50/60 Hz | |
50/60 Hz |
CE, CSA | |
<1 CFU/mL | |
Tap | |
15 Mohm.cm at 25°C | |
1 to 5 ppb | |
30 to 100 psig (2 to 6.9 bar) | |
<1 μM/mL | |
12 L/hr. | |
34.3 cm | |
49.5 cm | |
47 cm | |
100/240 VAC |