Learn More
Emma Safety Footwear BAS XD Safety Shoes
All purpose safety boot. The outsole repels acid and oil like an umbrella repels water. Strong ankle supporting upper fits the foot snugly and comfortably.
Brand:  Emma Safety Footwear MM70186841
Additional Details : Weight : 1.65100kg
 View more versions of this product
Product Code. 15918871
  • 122.96 € / 1 pair
Estimated Shipment: 07-10-2024
to see stock.
Add to basket
Description
Description
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction
  • GripForce™ Duo Heat Resistant Outsole
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Specifications
BAS XD
Leather
41