missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl-6 Mouse, Clone: BCL6/1527, enQuireBio™
Mouse Monoclonal Antibody
199.00 € - 485.00 €
Specifications
Antigen | Bcl-6 |
---|---|
Clone | BCL6/1527 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15930818
|
EnquireBio
604-MSM7-P1 |
100 μg |
485.00 €
100µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15920818
|
EnquireBio
604-MSM7-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Bcl-6 | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
604 | |
Recombinant human bcl-6 protein fragment (aa256-389) | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
95kDa |
BCL6/1527 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
NP_001124317 | |
BCL6 | |
IgG1 κ | |
Protein A/G Purified | |
Nuclear | |
Raji or Ramos cells. Tonsil or Hodgkin's lymphoma. |