Learn More
BD Antibody Diluent
For acetone-fixed, frozen tissue sections and formalin-fixed, paraffin-embedded tissue sections
Brand:  BD 559148
Additional Details : Weight : 0.12500kg
Product Code. 15879678
  • 173.00 € / Each
Estimated Shipment: 17-04-2024
to see stock.
Add to basket
Description
Description
Premixed, ready to use, sodium phosphate-based buffer solution, useful for the dilution of primary and/or secondary antibodies to be used in immunohistochemistry applications
  • Storage buffer: Aqueous buffered solution containing BSA, goat serum, and ≤0.09% sodium azide
Immunohistochemistry
Specifications
Specifications
Antibody diluent
Immunohistochemistry (IHC), Staining of Intra-cellular Antigens
4°C
125 mL
IHCH
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Research Use Only