Learn More
BD Difco™ YPD Broth
Used for maintaining and propagating yeasts. BD™ Difco™ YPD Broth is a dehydrated powder for preparing culture media in molecular microbiology procedures.
296.00 € - 545.00 €
Specifications
Color | Beige |
---|---|
Content And Storage | <30°C |
Form | Dehydrated Powder |
Media Type | YPD Broth |
Product Type | Microbiology Media |
Product Code | Brand | Packaging Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging Type | Quantity | Price | Quantity & Availability | ||||
11738882
|
BD
242810 |
Pail | 2 kg |
545.00 €
2kg |
Estimated Shipment: 03-07-2024
Log in to see stock. |
||||
10574579
|
BD
242820 |
Bottle | 500 g |
296.00 €
500g |
In Stock
Log in to see stock. |
||||
Description
General methods in yeast genetics specify using yeast extract-peptone-dextrose (YPD) medium for cultivating Saccharomyces cerevisiae and other yeasts. Yeasts grow well on a minimal medium containing only dextrose and salts. The addition of protein and yeast cell extract hydrolysates allows faster growth so that during exponential or log-phase growth, the cells divide every 90 minutes.
- Peptone acts as a source of carbon, nitrogen, vitamins and minerals
- Yeast extract supplies B-complex vitamins, which stimulate bacterial growth
- Dextrose is the carbohydrate source
- pH (6.5% solution, 25°C): 6.5 ± 0.2
Order Info
Prices are for units of one each unless otherwise specified.
Specifications
Beige | |
Dehydrated Powder | |
Microbiology Media | |
For maintaining and propagating Yeasts in molecular microbiology procedures |
<30°C | |
YPD Broth | |
Yeast extract 10g/L, Peptone 20g/L, Dextrose 20g/L |