missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Gelatin

Granulated gelatin for microbiological procedures.  BD Difco™ Gelatin is part of a large selection of dehydrated media.

Brand:  BD 214340

Additional Details : Weight : 0.99000kg

Product Code. 10566100

  • 168.00 € / 500g
Estimated Shipment: 04-06-2024
to see stock.
Add to basket

Description

Description

  • Use for: general microbiological procedures
  • Dehydrated
  • Culture media
  • Granulated gelatin
Specifications

Specifications

ISO/DIS, 11014
Gelatin
Culture media to detect Gelatin
500 g
<30°C
Dehydrated Granule
Bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers