Learn More
BD GasPak™ Supplies
Used with the GasPak™ EZ Gas Generating Systems. BD™ GasPak™ Supplies are used with these complete, integrated systems for rapid, waterless and catalyst-free gas generation. Can be used to support an anaerobic, microaerophilic or CO2 enriched environment.
Brand: BD 270303
Additional Details : Weight : 1.00000kg
Description
GasPak™ EZ Gas Generating Systems are complete, integrated systems that rapidly produce atmospheres suitable to support the primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria by use of gas generating sachets inside multi-use incubation containers.
Accessories include:
- CO2 Indicator Strips
- Dry Anaerobic Indicator Strips
- Tube Holder
- Catalyst Replacement Charges
- Catalyst Reaction Chamber
Specifications
Catalyst Replacement Charges | |
Produce atmospheres suitable to support the primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria | |
10 Pk. |
15°C to 30°C | |
BD GasPak™ |
Safety and Handling
- X10 SACHETS OF CATALYST (JAR22003)
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P391-Collect spillage.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : aluminium oxide