Learn More
BD NIH 3T3 Cell Lysate
Used as a control for the western blotting application
Brand: BD 611452
Additional Details : Weight : 0.01000kg
Includes: Mouse Embryonic Fibroblast.
Description
A source that can be rich in endogenously expressed proteins, this lysate is convenient to use and provided in a denatured format allowing for immediate loading onto an SDS-PAGE gel.
- Concentration: 1mg/mL
- Size: 500μg
Specifications
Embryonic Fibroblast | |
1mg/mL in SDS-PAGE Buffer: 62mM Tris pH 6.8, 2% SDS, 0.9% beta-mercaptoethanol, 0.003% Bromophenol Blue, 5% Glycerol | |
1 mg/mL |
500 μg | |
Mouse | |
-20°C |