missing translation for 'onlineSavingsMsg'
Learn More

Brand™ Low-Form Polypropylene Beakers with Blue Printed Scale

High clarity

Brand:  Brand™ 87626

Code : NEW

Additional Details : Weight : 1.52000kg

 View more versions of this product

Product Code. 15397864

  • 89.45 € / Pack of 4


Estimated Shipment
14-06-2019
Add to basket

Description

Description

  • With beaded rim and spout

Specifications

Specifications

Yes
500mL
5000 mL
Polypropylene
Videos