missing translation for 'onlineSavingsMsg'
Learn More

Brand™ Low-Form Polypropylene Beakers with Blue Printed Scale

High clarity

Brand:  Brand™ 87604

Code : NEW

Additional Details : Weight : 0.07000kg

 View more versions of this product

Product Code. 15397854

  • 15.72 € / Pack of 12


Estimated Shipment
14-06-2019
Add to basket

Description

Description

  • With beaded rim and spout

Specifications

Specifications

Yes
5mL
25 mL
Polypropylene
Videos