Learn More
Bel-Art™ SP Scienceware™ 1.5 mil Thick Biohazard Disposal Bags
Include black biohazard symbol and precautionary procedures printed in four languages. Bel-Art™ SP Scienceware™ 1.5 mil Thick Biohazard Disposal Bags are autoclavable and are effective for the disposal of biohazardous materials.
Brand: Bel-Art™ F13164-3848
Code : A0
Additional Details : Weight : 6.04800kg
Description
Include black biohazard symbol and precautionary procedures printed in four languages. Bel-Art™ SP Scienceware™ 1.5 mil Thick Biohazard Disposal Bags are autoclavable and are effective for the disposal of biohazardous materials.
- Bright red polypropylene bags include black biohazard symbol
- Precautionary procedures printed on bag in English, Spanish, French, and German for added safety
- Sterilization patch darkens when subjected to steam sterilization
- High temperature steam autoclavable to 135°C (275°F)
- Pass ASTM 1922 Tear Resistance and ASTM 1709 Dart Impact Tests
- Made in USA
Specifications
Biohazard Disposal Bag | |
Autoclavable | |
ASTM 1922 Tear Resistance, ASTM 1709 Dart Impact | |
Used for biohazard waste | |
Polypropylene | |
1.50 mil | |
97 cm | |
122 cm |
38W x 48H in. (97 x 122cm) | |
40 to 55 gal. | |
Red | |
Pre-printed sterilization indicator patch, labeled with the standard biohazard warning symbol or precautionary protocols English, French, German, Spanish | |
0.279 mm | |
38 in. | |
48 in. |