missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Filter Paper for Table-Top Buchner Funnels

Fitted specifically for Scienceware table-top Buchner funnels

POA

Specifications

Grade Heavy-bond
View More Specs

Description

Description

  • Made from creped white cellulose to improve porosity
  • Heavy-bond grade
  • Grammage value is 100g/sq.m
  • Thickness is 0.30mm
  • Flow rate is 90mL/minute
  • Wet burst strength is >68kPa
  • Particle Retention is 3μm

Specifications

Specifications

Heavy-bond
Videos
SDS
Documents

Documents

Product Certifications