missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Poxygrid™ Wire Test Tube Racks: 10-13mm
Chemical (organic solvents, salts, weak acids and weak alkalis) and corrosion-resistant steel wire racks are coated with heat-cured epoxy
40.90 € - 54.40 €
Specifications
Product Type | Test Tube Rack |
---|---|
Autoclavable | Autoclavable |
Holds | Tubes (10 to 13 mm) |
Material | Epoxy Coated Steel |
Diameter (Metric) Opening | 13 mm |
Product Code | Brand | Color | Positions | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Positions | Price | Quantity & Availability | ||||
15550012
|
Bel-Art™
F18750-0003 |
Orange | 72 |
42.30 €
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
||||
15520012
|
Bel-Art™
F18750-0000 |
Green | 72 |
42.90 €
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
||||
15530012
|
Bel-Art™
F18750-0001 |
Blue | 72 |
42.90 €
Each |
Estimated Shipment: 25-06-2024
Log in to see stock. |
||||
15560012
|
Bel-Art™
F18753-0001 |
Blue | 108 |
45.75 €
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
||||
13462499
|
Bel-Art™
F18751-0160 |
Blue | 60 |
40.90 €
Each |
Estimated Shipment: 20-06-2024
Log in to see stock. |
||||
13492499
|
Bel-Art™
F18752-1150 |
Blue | 150 |
51.20 €
Pack of 12 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||||
12962550
|
Bel-Art™
F18753-0000 |
Green | 108 |
54.40 €
Each |
Estimated Shipment: 25-06-2024
Log in to see stock. |
||||
15540012
|
Bel-Art™
F18750-0002 |
Yellow | 72 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Withstands temperature range of -223°C (-369°F) to 121°C (250°F)
- Consists of square holes, three decks, a slotted grid bottom and extra strength reinforced edges
- Orange racks can be used for biohazard warning while yellow for radioactive warning
- Epoxy coatings are unaffected by organic solvents, salts, weak acids and weak alkalis
- Stores from 40 to 108 tubes (five different diameters of testtubes)
- Autoclavable
Specifications
Test Tube Rack | |
Tubes (10 to 13 mm) | |
13 mm |
Autoclavable | |
Epoxy Coated Steel |