Learn More
Bel-Art™ SP Scienceware™ Replacement Chucks for Pipette Pump™ Pipettors
Grips pipets securely. Bel-Art™ SP Scienceware™ Replacement Chucks are silicone rubber grips used with Bel-Art™ Pipette Pumps for safe, accurate and trouble-free pipetting with simple, one-handed operation.
Brand:  Bel-Art™ H93789-0070
Code : NEW
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 15318750
  • 9.70 € / Each
Estimated Shipment: 18-06-2024
to see stock.
Add to basket
Description
Description
  • For use with Pipette Pump pipettor with thumbwheel and plunger aspiration
  • Pipettes fit smoothly into these flexible tapered chucks
  • Replacement chucks to fit various sizes/colors of Pipette Pumps in case of loss or damage
  • Silicone rubber is resistant to acids and alkalis
  • Disassemble parts for cleaning; chuck should not be autoclaved
Specifications
Specifications
Not Autoclavable
Silicone Rubber
Replacement Chucks
2mL pump pipettor