Learn More
Bel-Art™ SP Scienceware™ Spinbar™ Capsule Magnetic Stirring Bar
Capsule prevents stirring bar from spinning off center and damaging pH probes and gels.
Brand: Bel-Art™ F37152-0000
Code : A0
Additional Details : Weight : 0.80000kg
Description
- Polygon magnetic stirring bar spins within polystyrene capsule
- Prevents damage during pH adjustments and to gels during staining/destaining procedures
- Large, flat capsule keeps stirring bar consistently centered on bottom of vertical electrophoresis units to efficiently mix the running buffer
- Prevents stirring bar from being impeded by chemicals which, in water, transform into sticky gel and enables efficient dissolving
- After use, capsule can be easily disassembled for cleaning
- Autoclavable
Specifications
30 x 8mm polygon magnetic stirring bar | |
Polystyrene | |
Yes | |
Polygon |