missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Spinbar™ Magnetic Sink Strainer
Resistant to chemicals sink strainer has four strong magnets in each corner to attract and hold Spinbar magnetic stirring bars. Bel Art™ SP Scienceware™ Spinbar™ Magnetic Sink Strainer is a simple solution to prevent your Spinbar™ Magnetic Stirring Bars from going down the sink drain.
32.00 € - 36.70 €
Specifications
For Use With (Application) | Prevents stir bar from washing down drain |
---|---|
For Use With (Equipment) | Stirring bars |
Includes | 4 strong magnets in each corner, long slits in the sides and wide skirts |
Description
- Long slits in the sides and skirt allow fluids to drain easily and rapidly
- A wide skirt enables strainer to be used in a variety of sink drains; flat edge on one side allows it to be placed close to the sink wall
- Four legs on the underside anchor the strainer firmly in sink drain and prevent it from being dislodged by flowing liquid
- Color choices improve visibility of stirring bars
Specifications
Prevents stir bar from washing down drain | |
Stirring bars | |
4 strong magnets in each corner, long slits in the sides and wide skirts |