missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ifng, Rat, Biotin, Clone: XMG1.2, Abnova™
Rat monoclonal antibody raised against recombinant Ifng.
Brand: Abnova MAB5887.500ug
Additional Details : Weight : 0.00010kg
Specifications
Ifng | |
Monoclonal | |
Biotin | |
The optimal working dilution should be determined by the end user. | |
Ifng | |
Ifng | |
Recombinant protein corresponding to mouse Ifng. | |
RUO | |
15978 | |
Mouse | |
Liquid |
ELISA | |
XMG1.2 | |
Rat monoclonal antibody raised against recombinant Ifng. | |
In PBS (0.09% sodium azide) | |
IFN-g/IFN-gamma/Ifg | |
Rat | |
500 μg | |
Primary | |
Mouse Interferon-gamma (IFN-gamma)1-3. This is a neutralizing antibody. | |
Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 κ |