missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Stir and Add™ Cuvet Mixer
Made of clear acrylic with a straight handle, this tool is used when reactions are performed directly into cuvettes, to mix added reagents
57.45 €
Specifications
Diameter (English) | 0.125 in. |
---|---|
Diameter (Metric) | 3.17 mm |
Material | Acrylic |
Quantity | 36/Cs. |
For Use With (Application) | Used to add enzymes and other materials to solution in spectrophotometer cells, this tool is used when reactions are performed directly into cuvettes, to mix added reagents |
Product Code | Brand | Description | Length (English) | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Length (English) | Length (Metric) | Price | Quantity & Availability | |||
12338875
|
Bel-Art™
F37752-0000 |
Cell type: 10mm light path; Straight handle | 4.5 in. | 11.4 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10307814
|
Bel-Art™
F37752-0001 |
Cell type: Semi-micro; Straight handle | 5.1 in. | 13 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Up and down motion in cell provides complete mixing
- Shaft diameter 3.17mm (0.125 in.)
- Straight handle
Used to add enzymes and other materials to solution in spectrophotometer cells.
Specifications
0.125 in. | |
Acrylic | |
Used to add enzymes and other materials to solution in spectrophotometer cells, this tool is used when reactions are performed directly into cuvettes, to mix added reagents |
3.17 mm | |
36/Cs. |