Learn More
Bel-Art™ SP Scienceware™ Wide Mouth Color-Code Safety Labeled Wash Bottles
Easy-to-read labeling printed bottle promoting safe working environment
Brand: Bel-Art™ F11646-0612
Code : NEW
Additional Details : Weight : 0.50000kg
Includes: Labeling with Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols, Chemical Abstract Service (CAS) number, protective equipment symbols and target organ information
Description
- Wide mouth 53mm (2 in.) bottle for easy refilling
- Made from solvent-resistant low-density polyethylene
- Polypropylene closure with leakproof design
- Pictogram instructions promote universal safety regardless of setting
Use LYOB only with solvents compatible with LDPE.
Compliance
Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
Specifications
Not Autoclavable | |
53 mm | |
16 oz., 500 mL | |
Clear | |
LDPE |
Polypropylene | |
Wide | |
500 mL | |
Ethyl Acetate |