Learn More
Benserazide hydrochloride, Thermo Scientific™
Inhibitor of L-aromatic amino acid decarboxylase
77.70 € - 239.00 €
Chemical Identifiers
| CAS | 14919-77-8 |
|---|---|
| Molecular Formula | C10H16ClN3O5 |
| Molecular Weight (g/mol) | 293.704 |
| MDL Number | MFCD00078571 |
| InChI Key | ULFCBIUXQQYDEI-UHFFFAOYSA-N |
| Synonym | benserazide hydrochloride, benserazide hcl, 2-amino-3-hydroxy-n'-2,3,4-trihydroxybenzyl propanehydrazide hydrochloride, ccris 5092, 2'-2,3,4-trihydroxybenzyl-dl-serinohydrazide monohydrochloride, benzerazide hydrochloride, dl-serine 2, dl-serine 2-2,3,4-trihydroxybenzyl hydrazine hydrochloride, 2-amino-3-hydroxy-n'-2,3,4-trihydroxyphenyl methyl propanehydrazide hydrochloride |
| PubChem CID | 26964 |
| ChEBI | CHEBI:31262 |
| IUPAC Name | 2-amino-3-hydroxy-N'-[(2,3,4-trihydroxyphenyl)methyl]propanehydrazide;hydrochloride |
| SMILES | C1=CC(=C(C(=C1CNNC(=O)C(CO)N)O)O)O.Cl |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15486469
|
Thermo Scientific Alfa Aesar
J64581.03 |
1g |
77.70 €
1g |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
15496469
|
Thermo Scientific Alfa Aesar
J64581.06 |
5g |
239.00 €
5g |
Estimated Shipment: 28-09-2026
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsInhibitor of L-aromatic amino acid decarboxylaseBenserazide hydrochloride is used as an antiparkinsonian. It is a peripheral decarboxylase inhibitor. It is used as an inhibitor for aromatic L-aminoacid decarboxylase. Further, it serves as an inhibitor of tyrosine hydroxylase (TH) and Tryptophan hydroxylase (TPH) inhibitor.
Solubility
Soluble in water, dimethyl sulfoxide and methanol. Insoluble in ethanol and acetone.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
| 14919-77-8 | |
| 293.704 | |
| ULFCBIUXQQYDEI-UHFFFAOYSA-N | |
| 26964 | |
| 2-amino-3-hydroxy-N'-[(2,3,4-trihydroxyphenyl)methyl]propanehydrazide;hydrochloride |
| C10H16ClN3O5 | |
| MFCD00078571 | |
| benserazide hydrochloride, benserazide hcl, 2-amino-3-hydroxy-n'-2,3,4-trihydroxybenzyl propanehydrazide hydrochloride, ccris 5092, 2'-2,3,4-trihydroxybenzyl-dl-serinohydrazide monohydrochloride, benzerazide hydrochloride, dl-serine 2, dl-serine 2-2,3,4-trihydroxybenzyl hydrazine hydrochloride, 2-amino-3-hydroxy-n'-2,3,4-trihydroxyphenyl methyl propanehydrazide hydrochloride | |
| CHEBI:31262 | |
| C1=CC(=C(C(=C1CNNC(=O)C(CO)N)O)O)O.Cl |
Specifications
| 14919-77-8 | |
| MFCD00078571 | |
| 14,1050 | |
| Soluble in water,dimethyl sulfoxide and methanol. Insoluble in ethanol and acetone. | |
| C1=CC(=C(C(=C1CNNC(=O)C(CO)N)O)O)O.Cl | |
| 293.704 | |
| CHEBI:31262 | |
| Benserazide hydrochloride |
| C10H16ClN3O5 | |
| Light sensitive | |
| benserazide hydrochloride, benserazide hcl, 2-amino-3-hydroxy-n'-2,3,4-trihydroxybenzyl propanehydrazide hydrochloride, ccris 5092, 2'-2,3,4-trihydroxybenzyl-dl-serinohydrazide monohydrochloride, benzerazide hydrochloride, dl-serine 2, dl-serine 2-2,3,4-trihydroxybenzyl hydrazine hydrochloride, 2-amino-3-hydroxy-n'-2,3,4-trihydroxyphenyl methyl propanehydrazide hydrochloride | |
| ULFCBIUXQQYDEI-UHFFFAOYSA-N | |
| 2-amino-3-hydroxy-N'-[(2,3,4-trihydroxyphenyl)methyl]propanehydrazide;hydrochloride | |
| 26964 | |
| 293.7 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 238-991-9
missing translation for 'rtecsNumber' : VT9632300
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.