missing translation for 'onlineSavingsMsg'
Learn More

Benzalkonium chloride, alkyl distribution C8-C16, 50 wt% aqueous solution, Thermo Scientific Chemicals

Catalog Number p-3625870
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
8001-54-5, 7732-18-5
C9H13ClN
170.66
MFCD00145757
Alkyl-benzyl-dimethylammonium chloride
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
8001-54-5, 7732-18-5
C9H13ClN
170.66
MFCD00145757
Alkyl-benzyl-dimethylammonium chloride

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Boiling Point >100.0°C
CAS Min % 49.0, 48.0
CAS Max % 52.0, 51.0
Linear Formula C6H5CH2N(CH3)2RClR=C8H17toC18H37
Concentration 49 to 52% (based on M.W.= 352.5)
Density 0.9800g/mL
Merck Index 15, 1061
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and acetone, slightly soluble in benzene, almost insoluble in ether
Packaging Glass bottle
Specific Gravity 0.98
Viscosity 130 mPa.s (20°C)
pH 6 to 9 (10% aq. soln.)
Quantity 1 L
Flash Point 105°C
Physical Form Solution
Chemical Name or Material Benzalkonium chloride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Benzalkonium chloride, 50 wt% aqueous solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'