missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzethonium chloride, 97%, Thermo Scientific Chemicals
43.40 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Melting Point | 158.0°C to 163.0°C |
Color | White |
Loss on Drying | 5% max. (105°C, 4 hrs) |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (on dry substance) (Argentometry) |
Packaging | Plastic bottle |
Quantity | 100 g |
Fieser | 01,433 |
Show More |
Safety and Handling
Product Identifier
- Benzethonium chloride
- Danger
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 C
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
- H301-Toxic if swallowed.
- H314-Causes severe skin burns and eye damage.
- H400-Very toxic to aquatic life.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P273-Avoid release to the environment.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'