missing translation for 'onlineSavingsMsg'
Learn More

Benzil, 99+%, Thermo Scientific Chemicals

Catalog Number 11964841
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
134-81-6
C14H10O2
210.23
MFCD00003080
WURBFLDFSFBTLW-UHFFFAOYSA-N
benzil, diphenylethanedione, dibenzoyl, diphenylglyoxal, bibenzoyl, ethanedione, diphenyl, 1,2-diphenylethanedione, diphenylethane-1,2-dione, glyoxal, diphenyl, diphenyldiketon
8651
CHEBI:51507
1,2-diphenylethane-1,2-dione
O=C(C(=O)C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
134-81-6
C14H10O2
210.23
MFCD00003080
WURBFLDFSFBTLW-UHFFFAOYSA-N
benzil, diphenylethanedione, dibenzoyl, diphenylglyoxal, bibenzoyl, ethanedione, diphenyl, 1,2-diphenylethanedione, diphenylethane-1,2-dione, glyoxal, diphenyl, diphenyldiketon
8651
CHEBI:51507
1,2-diphenylethane-1,2-dione
O=C(C(=O)C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point 94.0°C to 97.0°C
Color White
Boiling Point 346.0°C to 348.0°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Linear Formula C6H5COCOC6H5
Quantity 2.5 kg
Merck Index 15,108
Solubility Information Solubility in water: 0.5g/L (20°C). Other solubilities: soluble in alcohol,ether,chloroform,ethyl,acetate,benzene,toluene and nitrobenzene
Formula Weight 210.23
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Benzil, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • Benzil
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'